Learn More
Miele, Inc. E 144 Half Insert for beakers (18 beakers, up to 250 ml)
Catalog No. NC1330087
Encompass_Preferred
E 144 Mesh Basket for the optimum loading of up to 18 beakers.
Suitable for 18 glass beakers
Equipped with 18 x 3 stay bars
For use in upper and lower basket
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC1330087 Supplier Miele, Inc. Supplier No. 03808710
E 144 Mesh Basket for the optimum loading of up to 18 beakers.
Suitable for 18 glass beakers
Equipped with 18 x 3 stay bars
For use in upper and lower basket
Visit www.mieleusa.com/professional for technical information.