Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-2-[2-[2-[5-[4-[tert-butoxycarbonyl(methyl)amino]styryl]pyridine-2-yloxy]ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate | 1205550-99-7 | 99.7% | 10 MG
SDP

Catalog No. 5000232936
Encompass_Preferred

AV-105 is a tosylate precursor used in the synthesis of florbetapir (18F), a PET imaging tracer. It is supplied as a solid with high reported purity for radiochemistry and tracer development.

  • Used as precursor for florbetapir (18F) synthesis.
  • Supplied as a solid with high reported purity (99.7%).
  • Molecular weight 612.73 g/mol.
  • Chemical formula C32H40N2O8S.
  • Packaged in 10 MG quantities for research use.

Catalog No. 50-002-32936 Supplier Medchemexpress LLC Supplier No. HY12094710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AV-105 is a tosylate precursor used in the synthesis of florbetapir (18F), a PET imaging tracer. It is supplied as a solid with high reported purity for radiochemistry and tracer development.

  • Used as precursor for florbetapir (18F) synthesis.
  • Supplied as a solid with high reported purity (99.7%).
  • Molecular weight 612.73 g/mol.
  • Chemical formula C32H40N2O8S.
  • Packaged in 10 MG quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.