Learn More
Medchemexpress LLC (E)-2,3',4,5'-tetramethoxystilbene | 24144-92-1 | 99.4% | 300.35 | 1 ML
SDP
Catalog No. 5000385986
Encompass_Preferred
TMS ((E)-2,3′,4,5′-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. It shows a lesser inhibitory effect on CYP1A1 (IC50=300 nM) and CYP1A2 (IC50=3.1 μM). This methylated derivative of resveratrol possesses anti-cancer activity.
  • Selective and competitive CYP1B1 inhibitor
  • Possesses anti-cancer activity
  • IC50 for CYP1B1 is 6 nM
  • IC50 for CYP1A1 is 300 nM
  • IC50 for CYP1A2 is 3.1 μM
Catalog No. 50-003-85986 Supplier Medchemexpress LLC Supplier No. HY1934010MM/1ML
Add to Cart
Add to Cart
TMS ((E)-2,3′,4,5′-tetramethoxystilbene) is a selective and competitive CYP1B1 inhibitor with an IC50 of 6 nM and a Ki value of 3 nM. It shows a lesser inhibitory effect on CYP1A1 (IC50=300 nM) and CYP1A2 (IC50=3.1 μM). This methylated derivative of resveratrol possesses anti-cancer activity.
  • Selective and competitive CYP1B1 inhibitor
  • Possesses anti-cancer activity
  • IC50 for CYP1B1 is 6 nM
  • IC50 for CYP1A1 is 300 nM
  • IC50 for CYP1A2 is 3.1 μM