Learn More
Medchemexpress LLC (E)-2-(3,4-dihydroxybenzoyl)-3-(2-oxo-3H-1,3-benzoxazol-5-yl)prop-2-enenitrile | 638213-98-6 | 98.3% | 322.27 | 10 MM 1 ML
SDP
Catalog No. 5000437454
Encompass_Preferred
AGL-2263 is a novel and potent inhibitor of both the insulin receptor (IR) and the insulin-like growth factor (IGF) receptor. It appears as a light yellow to yellow solid powder and is intended for research use only, not for human or veterinary use.
  • Inhibits the insulin receptor (IR) and insulin-like growth factor (IGF) receptor.
  • Available as a light yellow to yellow solid powder.
  • Stable at ambient temperature for ordinary shipping.
  • Soluble in DMSO (~100 mg/mL) and various formulations.
  • Demonstrates biological activity in vitro by inhibiting IR.
  • Can still allow insulin to phosphorylate ERK1/2 and AKT in some cellular contexts.
Catalog No. 50-004-37454 Supplier Medchemexpress LLC Supplier No. HY11272010MM/1ML
AGL-2263 is a novel and potent inhibitor of both the insulin receptor (IR) and the insulin-like growth factor (IGF) receptor. It appears as a light yellow to yellow solid powder and is intended for research use only, not for human or veterinary use.
  • Inhibits the insulin receptor (IR) and insulin-like growth factor (IGF) receptor.
  • Available as a light yellow to yellow solid powder.
  • Stable at ambient temperature for ordinary shipping.
  • Soluble in DMSO (~100 mg/mL) and various formulations.
  • Demonstrates biological activity in vitro by inhibiting IR.
  • Can still allow insulin to phosphorylate ERK1/2 and AKT in some cellular contexts.