Learn More
Medchemexpress LLC (E)-2-(3,4-dihydroxybenzoyl)-3-(4-hydroxy-3-iodo-5-methoxyphenyl)prop-2-enenitrile | 1094048-77-7 | MFCD03453582 | 98.0% | C17H12INO5 | 10MG
SDP
Catalog No. 5000259001
Encompass_Preferred
I-OMe-Tyrphostin AG 538 is a small-molecule inhibitor of the insulin-like growth factor-1 receptor (IGF-1R) and PI5P4Kα used in biochemical and cellular research to probe IGF-1R-mediated signaling. Supplied as a high-purity solid for in vitro studies.
  • Specific inhibitor of IGF-1R and PI5P4Kα.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Solid, yellow to orange appearance for easy handling.
  • Molecular formula C17H12INO5; molecular weight 437.19.
  • Stable under recommended storage conditions for reliable results.
  • Provided in small research quantities for laboratory experiments.
Catalog No. 50-002-59001 Supplier Medchemexpress LLC Supplier No. HY13568010MG
I-OMe-Tyrphostin AG 538 is a small-molecule inhibitor of the insulin-like growth factor-1 receptor (IGF-1R) and PI5P4Kα used in biochemical and cellular research to probe IGF-1R-mediated signaling. Supplied as a high-purity solid for in vitro studies.
  • Specific inhibitor of IGF-1R and PI5P4Kα.
  • High purity (98.0%) suitable for biochemical and cellular assays.
  • Solid, yellow to orange appearance for easy handling.
  • Molecular formula C17H12INO5; molecular weight 437.19.
  • Stable under recommended storage conditions for reliable results.
  • Provided in small research quantities for laboratory experiments.