Learn More
Medchemexpress LLC (E)-2-(((5-bromothiophen-2-yl)methylene)amino)benzamide | 304456-62-0 | 98.0% | 309.18 | C12H9BrN2OS | 5MG
SDP
Catalog No. 5000237430
Encompass_Preferred
BTYNB is a small-molecule research inhibitor of IMP1 (IGF2BP1) that disrupts IMP1 binding to c-Myc mRNA, resulting in reduced c-Myc expression and decreased proliferation in IMP1-positive cancer cell lines. The compound is supplied as a solid reagent in multiple sizes and as a 10 mM solution in DMSO for convenient stock preparation, and is reported with high purity for in vitro applications.
  • Inhibits IMP1 binding to c-Myc mRNA with reported IC50 ≈ 5 μM.
  • High reported purity suitable for in vitro research applications.
  • Available in multiple solid pack sizes and a 10 mM solution format in DMSO.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Used to study RNA-protein interactions and cancer cell proliferation.
Catalog No. 50-002-37430 Supplier Medchemexpress LLC Supplier No. HY1244475MG
BTYNB is a small-molecule research inhibitor of IMP1 (IGF2BP1) that disrupts IMP1 binding to c-Myc mRNA, resulting in reduced c-Myc expression and decreased proliferation in IMP1-positive cancer cell lines. The compound is supplied as a solid reagent in multiple sizes and as a 10 mM solution in DMSO for convenient stock preparation, and is reported with high purity for in vitro applications.
  • Inhibits IMP1 binding to c-Myc mRNA with reported IC50 ≈ 5 μM.
  • High reported purity suitable for in vitro research applications.
  • Available in multiple solid pack sizes and a 10 mM solution format in DMSO.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Used to study RNA-protein interactions and cancer cell proliferation.