Learn More
Medchemexpress LLC (E)-2-amino-4-methyl-5-phosphonopent-3-enoic acid | 127910-31-0 | MFCD00153906 | 98.3% | 209.14 g·mol-1 | C6H12NO5P | 1 MG
SDP
Catalog No. 5000240743
Encompass_Preferred
CGP 37849 is a selective, competitive N-methyl-D-aspartate (NMDA) receptor antagonist with reported oral activity. It has demonstrated anticonvulsant activity and antidepressant- and anxiolytic-like effects in rodent models, and is used in pharmacology research to investigate NMDA-mediated neurotransmission.
  • Selective competitive NMDA receptor antagonist
  • Orally active with demonstrated in vivo anticonvulsant effects
  • Shows antidepressant- and anxiolytic-like effects in rodent studies
  • Chemical formula: C6H12NO5P; molecular weight: 209.14 g·mol-1
  • Supplied in small milligram quantities suitable for research use
Catalog No. 50-002-40743 Supplier Medchemexpress LLC Supplier No. HY1077021MG
Add to Cart
Add to Cart
CGP 37849 is a selective, competitive N-methyl-D-aspartate (NMDA) receptor antagonist with reported oral activity. It has demonstrated anticonvulsant activity and antidepressant- and anxiolytic-like effects in rodent models, and is used in pharmacology research to investigate NMDA-mediated neurotransmission.
  • Selective competitive NMDA receptor antagonist
  • Orally active with demonstrated in vivo anticonvulsant effects
  • Shows antidepressant- and anxiolytic-like effects in rodent studies
  • Chemical formula: C6H12NO5P; molecular weight: 209.14 g·mol-1
  • Supplied in small milligram quantities suitable for research use