Learn More
Medchemexpress LLC (E)-2-cyano-3-(3,4-dihydroxyphenyl)-N-[(1S)-1-phenylethyl]prop-2-enamide | 133550-37-5 | MFCD00209855 | 97.0% | 308.33 g/mol | C18H16N2O3 | 1 ML
SDP
Catalog No. 5000211898
Encompass_Preferred
(+)-Tyrphostin B44 is the (+) enantiomer of Tyrphostin AG-835, a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) used in biochemical and cellular research. It is provided as a concentrated solution in DMSO for in vitro assays and is documented with supporting datasheet, certificate of analysis, and safety data information.
  • Provides EGFR inhibitory activity useful for signaling and cancer research.
  • Supplied as a 10 mM solution in DMSO for ready-to-use preparations.
  • High purity suitable for research applications (≈97%).
  • Characterized molecular properties for accurate dosing in assays.
  • Documentation available to support safe handling and experimental reproducibility.
Catalog No. 50-002-11898 Supplier Medchemexpress LLC Supplier No. HY101007A10MM 1ML
(+)-Tyrphostin B44 is the (+) enantiomer of Tyrphostin AG-835, a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) used in biochemical and cellular research. It is provided as a concentrated solution in DMSO for in vitro assays and is documented with supporting datasheet, certificate of analysis, and safety data information.
  • Provides EGFR inhibitory activity useful for signaling and cancer research.
  • Supplied as a 10 mM solution in DMSO for ready-to-use preparations.
  • High purity suitable for research applications (≈97%).
  • Characterized molecular properties for accurate dosing in assays.
  • Documentation available to support safe handling and experimental reproducibility.