Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(4-phenylbutyl)-2-propenamide | 133550-41-1 | 99.9% | 336.39 g/mol | C20H20N2O3 | 1 ML
SDP

Catalog No. 5000212087
Encompass_Preferred

(E)-AG 556 is a small-molecule, selective inhibitor of the epidermal growth factor receptor (EGFR) used in preclinical research to probe EGFR-mediated signaling and inflammatory responses. It inhibits EGFR kinase activity and has been reported to reduce LPS-induced TNF-α production. The compound is supplied as a high-purity powder and is also available as a ready-to-use 10 mM stock solution in 1 mL vials for convenient dosing in cellular and biochemical assays.

  • Selective EGFR inhibitor for signaling studies.
  • Reported to block LPS-induced TNF-α production in cell assays.
  • High purity suitable for biochemical and cell-based assays.
  • Available as powder and as a 10 mM stock solution for immediate use.
  • Molecular formula C20H20N2O3 and molecular weight 336.39 g/mol.

Catalog No. 50-002-12087 Supplier Medchemexpress LLC Supplier No. HY10104110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-AG 556 is a small-molecule, selective inhibitor of the epidermal growth factor receptor (EGFR) used in preclinical research to probe EGFR-mediated signaling and inflammatory responses. It inhibits EGFR kinase activity and has been reported to reduce LPS-induced TNF-α production. The compound is supplied as a high-purity powder and is also available as a ready-to-use 10 mM stock solution in 1 mL vials for convenient dosing in cellular and biochemical assays.

  • Selective EGFR inhibitor for signaling studies.
  • Reported to block LPS-induced TNF-α production in cell assays.
  • High purity suitable for biochemical and cell-based assays.
  • Available as powder and as a 10 mM stock solution for immediate use.
  • Molecular formula C20H20N2O3 and molecular weight 336.39 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.