Learn More
Medchemexpress LLC (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(4-phenylbutyl)-2-propenamide | 133550-41-1 | 99.9% | 336.39 g/mol | C20H20N2O3 | 10 MG
SDP
Catalog No. 5000213089
Encompass_Preferred
(E)-AG 556 is a research-grade small-molecule inhibitor of the epidermal growth factor receptor (EGFR). It is highly selective for EGFR kinase activity and has been reported to block lipopolysaccharide (LPS)-induced tumor necrosis factor-alpha (TNF-α) production. The compound is supplied as a solid with high reported purity and is typically dissolved in DMSO for in vitro studies.
  • Highly selective EGFR tyrosine kinase inhibitor.
  • Reported to block LPS-induced TNF-α production.
  • High purity (reported ~99.9%).
  • Solid, light yellow to yellow appearance.
  • Soluble in DMSO for preparation of stock solutions.
  • Suitable for cell-based and biochemical assays.
Catalog No. 50-002-13089 Supplier Medchemexpress LLC Supplier No. HY10104110MG
(E)-AG 556 is a research-grade small-molecule inhibitor of the epidermal growth factor receptor (EGFR). It is highly selective for EGFR kinase activity and has been reported to block lipopolysaccharide (LPS)-induced tumor necrosis factor-alpha (TNF-α) production. The compound is supplied as a solid with high reported purity and is typically dissolved in DMSO for in vitro studies.
  • Highly selective EGFR tyrosine kinase inhibitor.
  • Reported to block LPS-induced TNF-α production.
  • High purity (reported ~99.9%).
  • Solid, light yellow to yellow appearance.
  • Soluble in DMSO for preparation of stock solutions.
  • Suitable for cell-based and biochemical assays.