Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoic acid | 122520-79-0 | MFCD09739079 | 99.7% | 205.17 g·mol⁻¹ | C10H7NO4 | 2 MG
SDP

Catalog No. 5000227608
Encompass_Preferred

Tyrphostin AG30 is a small-molecule EGFR tyrosine kinase inhibitor (CAS 122520-79-0) used in biochemical and cellular studies to probe EGFR/c-ErbB signaling. It is supplied as a high-purity solid appropriate for solution preparation and in vitro testing.

  • High purity: 99.7%.
  • Target: EGFR tyrosine kinase; inhibits c-ErbB-induced self-renewal and STAT5 activation.
  • Molecular formula C10H7NO4 and molecular weight 205.17 g·mol⁻¹.
  • Available in milligram pack sizes for small-scale research use.
  • Storage: powder -20 °C (long-term) or 4 °C (short-term); in solvent -80 °C recommended for extended stability.

Catalog No. 50-002-27608 Supplier Medchemexpress LLC Supplier No. HY1185322MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG30 is a small-molecule EGFR tyrosine kinase inhibitor (CAS 122520-79-0) used in biochemical and cellular studies to probe EGFR/c-ErbB signaling. It is supplied as a high-purity solid appropriate for solution preparation and in vitro testing.

  • High purity: 99.7%.
  • Target: EGFR tyrosine kinase; inhibits c-ErbB-induced self-renewal and STAT5 activation.
  • Molecular formula C10H7NO4 and molecular weight 205.17 g·mol⁻¹.
  • Available in milligram pack sizes for small-scale research use.
  • Storage: powder -20 °C (long-term) or 4 °C (short-term); in solvent -80 °C recommended for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.