Learn More
Medchemexpress LLC (E)-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enoic acid | 122520-79-0 | MFCD09739079 | 99.7% | 205.17 g/mol | C10H7NO4 | 25 MG
SDP
Catalog No. 5000227477
Encompass_Preferred
Tyrphostin AG30 is a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) tyrosine kinase used in biochemical and cellular research to probe EGFR-mediated signaling. The compound is a light yellow solid (C10H7NO4, MW 205.17 g/mol) supplied at high purity for reproducible assay results, with recommended cold storage for long-term stability.
  • Potent EGFR tyrosine kinase inhibitor.
  • High chemical purity for consistent experimental data.
  • Light yellow solid suitable for formulation and assay preparation.
  • Recommended cold storage for long-term stability.
  • Available in research-scale quantities for biochemical and cellular studies.
Catalog No. 50-002-27477 Supplier Medchemexpress LLC Supplier No. HY11853225MG
Tyrphostin AG30 is a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) tyrosine kinase used in biochemical and cellular research to probe EGFR-mediated signaling. The compound is a light yellow solid (C10H7NO4, MW 205.17 g/mol) supplied at high purity for reproducible assay results, with recommended cold storage for long-term stability.
  • Potent EGFR tyrosine kinase inhibitor.
  • High chemical purity for consistent experimental data.
  • Light yellow solid suitable for formulation and assay preparation.
  • Recommended cold storage for long-term stability.
  • Available in research-scale quantities for biochemical and cellular studies.