Learn More
Medchemexpress LLC (E)-2-Hydroxycinnamaldehyde | 60125-23-7 | 1 ML
SDP
Catalog No. 5000421749
Encompass_Preferred
(E)-2-Hydroxycinnamaldehyde is an aldehyde that can be separated from the stem bark of cinnamon. It acts as an antitumor agent by inhibiting cell proliferation and inducing apoptosis. This is achieved by inhibiting the signal transduction of STAT3 and the production of reactive oxygen species. It is for research use only and not sold to patients.
  • Inhibits cell proliferation
  • Induces apoptosis by inhibiting signal transduction of STAT3
  • Inhibits reactive oxygen species production
  • Has antitumor activity
  • For research use only
Catalog No. 50-004-21749 Supplier Medchemexpress LLC Supplier No. HYN314910MM/1ML
Add to Cart
Add to Cart
(E)-2-Hydroxycinnamaldehyde is an aldehyde that can be separated from the stem bark of cinnamon. It acts as an antitumor agent by inhibiting cell proliferation and inducing apoptosis. This is achieved by inhibiting the signal transduction of STAT3 and the production of reactive oxygen species. It is for research use only and not sold to patients.
  • Inhibits cell proliferation
  • Induces apoptosis by inhibiting signal transduction of STAT3
  • Inhibits reactive oxygen species production
  • Has antitumor activity
  • For research use only