Learn More
Medchemexpress LLC (E)-2-Hydroxycinnamaldehyde | 60125-23-7 | 250 MG
SDP
Catalog No. 5000422072
Encompass_Preferred
(E)-2-Hydroxycinnamaldehyde is an aldehyde that can be isolated from the stem bark of cinnamon. It functions by inhibiting cell proliferation and inducing apoptosis through the suppression of STAT3 signal transduction and reactive oxygen species production. This compound exhibits antitumor activity.
  • Inhibits cell proliferation
  • Induces apoptosis by suppressing STAT3 signal transduction
  • Induces apoptosis by suppressing reactive oxygen species production
  • Exhibits antitumor activity
Catalog No. 50-004-22072 Supplier Medchemexpress LLC Supplier No. 250MGHYN3149
(E)-2-Hydroxycinnamaldehyde is an aldehyde that can be isolated from the stem bark of cinnamon. It functions by inhibiting cell proliferation and inducing apoptosis through the suppression of STAT3 signal transduction and reactive oxygen species production. This compound exhibits antitumor activity.
  • Inhibits cell proliferation
  • Induces apoptosis by suppressing STAT3 signal transduction
  • Induces apoptosis by suppressing reactive oxygen species production
  • Exhibits antitumor activity