Learn More
Grainger AMS E-280 Pocket Penetrometer, Anodized Aluminum, 1 ea
Catalog No. 502791883
Encompass_Preferred
The pocket penetrometer is a spring-operated instrument designed for accurate measurement of compressive soil strength in cohesive soils. It is essential for field assessments to help reduce human error in soil classification and is mandated for use on all excavation jobs by OSHA construction standards. The unit features an anodized aluminum construction with a loading piston that provides readings in tons/sq. ft. or kg/sq. cm.
  • Provides accurate measurements of compressive soil strength
  • Includes a friction ring to maintain maximum reading until manually reset
  • Features a 0.25 in. diameter loading piston with a calibration groove at 0.25 in. depth
  • Includes an Allen wrench and plastic carrying case with belt loop
  • Compatible with torsional vane shear testers for rapid soil friction estimation
  • Offers a measurement scale range of 0 to 4.5 tons/sq. ft.
Catalog No. 50-279-1883 Supplier Grainger Supplier No. 16D171
The pocket penetrometer is a spring-operated instrument designed for accurate measurement of compressive soil strength in cohesive soils. It is essential for field assessments to help reduce human error in soil classification and is mandated for use on all excavation jobs by OSHA construction standards. The unit features an anodized aluminum construction with a loading piston that provides readings in tons/sq. ft. or kg/sq. cm.
  • Provides accurate measurements of compressive soil strength
  • Includes a friction ring to maintain maximum reading until manually reset
  • Features a 0.25 in. diameter loading piston with a calibration groove at 0.25 in. depth
  • Includes an Allen wrench and plastic carrying case with belt loop
  • Compatible with torsional vane shear testers for rapid soil friction estimation
  • Offers a measurement scale range of 0 to 4.5 tons/sq. ft.