Learn More
Medchemexpress LLC [(E,2S,3R)-2-(dodecanoylamino)-3-hydroxyoctadec-4-enyl] 2-(trimethylazaniumyl)ethyl phosphate | 474923-21-2 | MFCD22416889 | 99.0% | 646.92 g/mol | C35H71N2O6P | 1mg
SDP
Catalog No. 5000269613
Encompass_Preferred
Sphingomyelin (d18 1/12 0) is a polar lipid with the activity of regulating cell signaling Sphingomyelin (d18 1/12 0) plays an important role in cell membranes affecting membrane fluidity and stability Sphingomyelin (d18 1/12 0) is also involved in lipid metabolism and has the potential to promote neuronal cell growth Sphingomyelin (d18 1/12 0) plays a key role in the function and development of the nervous system
Catalog No. 50-002-69613 Supplier Medchemexpress LLC Supplier No. HYW9236421MG
Sphingomyelin (d18 1/12 0) is a polar lipid with the activity of regulating cell signaling Sphingomyelin (d18 1/12 0) plays an important role in cell membranes affecting membrane fluidity and stability Sphingomyelin (d18 1/12 0) is also involved in lipid metabolism and has the potential to promote neuronal cell growth Sphingomyelin (d18 1/12 0) plays a key role in the function and development of the nervous system