Learn More
Medchemexpress LLC (E)-3-((1H-indol-3-yl)methylene)-5-((E)-3,4-dimethoxybenzylidene)-1-methylpiperidin-4-one | 1370032-19-1 | 99.1% | 388.46 g/mol | C24H24N2O3 | 5 MG
SDP
Catalog No. 5000262965
Encompass_Preferred
CA-5f is a small-molecule late-stage macroautophagy inhibitor that blocks autophagosome-lysosome fusion, elevates LC3B-II and SQSTM1/p62, increases reactive oxygen species, and has reported antitumor activity in cellular studies. Supplied for research use in solid form and as ready-to-use DMSO solutions; documentation is available for quality control.
  • Blocks autophagosome-lysosome fusion in autophagy assays.
  • Modulates LC3B-II and SQSTM1/p62 levels for pathway studies.
  • Induces reactive oxygen species in cell-based assays.
  • Available as solid and DMSO solution formats for flexibility.
  • High reported purity suitable for biochemical and cellular experiments.
  • Documentation includes datasheet, COA, and SDS for quality control.
Catalog No. 50-002-62965 Supplier Medchemexpress LLC Supplier No. HY1126985MG
CA-5f is a small-molecule late-stage macroautophagy inhibitor that blocks autophagosome-lysosome fusion, elevates LC3B-II and SQSTM1/p62, increases reactive oxygen species, and has reported antitumor activity in cellular studies. Supplied for research use in solid form and as ready-to-use DMSO solutions; documentation is available for quality control.
  • Blocks autophagosome-lysosome fusion in autophagy assays.
  • Modulates LC3B-II and SQSTM1/p62 levels for pathway studies.
  • Induces reactive oxygen species in cell-based assays.
  • Available as solid and DMSO solution formats for flexibility.
  • High reported purity suitable for biochemical and cellular experiments.
  • Documentation includes datasheet, COA, and SDS for quality control.