Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3-(2-((4-chlorophenyl)thio)phenyl)-N-(4-(dimethylamino)butyl)acrylamide hydrochloride | 1217201-17-6 | >99.0% | 425.41 | C21H25ClN2OS·HCl | 5 MG
SDP

Catalog No. 5000240383
Encompass_Preferred

A-350619 hydrochloride is a research-grade small-molecule activator of soluble guanylate cyclase (sGC). Supplied as the hydrochloride salt with high purity, it is used in preclinical studies probing NO-sGC signalling pathways and pharmacology related to erectile dysfunction.

  • Activates soluble guanylate cyclase (sGC) in biochemical and cellular assays.
  • Supplied as the hydrochloride salt for improved solubility.
  • High purity suitable for research use (>99% HPLC).
  • Molecular weight 425.41.
  • Store at 4 °C to maintain stability.
  • Applicable to preclinical studies of NO-sGC signalling and erectile dysfunction models.

Catalog No. 50-002-40383 Supplier Medchemexpress LLC Supplier No. HY1075485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A-350619 hydrochloride is a research-grade small-molecule activator of soluble guanylate cyclase (sGC). Supplied as the hydrochloride salt with high purity, it is used in preclinical studies probing NO-sGC signalling pathways and pharmacology related to erectile dysfunction.

  • Activates soluble guanylate cyclase (sGC) in biochemical and cellular assays.
  • Supplied as the hydrochloride salt for improved solubility.
  • High purity suitable for research use (>99% HPLC).
  • Molecular weight 425.41.
  • Store at 4 °C to maintain stability.
  • Applicable to preclinical studies of NO-sGC signalling and erectile dysfunction models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.