Learn More
Medchemexpress LLC (E)-3-(3-(2-chlorophenyl)acryloyl)-2H-chromen-2-one | 1253978-24-3 | 99.1% | 310.73 g/mol | C18H11ClO3 | 5 MG
SDP
Catalog No. 5000263156
Encompass_Preferred
MAO-B-IN-2 is a small-molecule, selective and competitive inhibitor of monoamine oxidase B (MAO-B) and butyrylcholinesterase (BChE). It is provided for research use and is available as a solid and as a 10 mM solution in DMSO for biochemical and cell-based studies.
  • Selective MAO-B and BChE inhibition (IC50 MAO-B = 0.51 μM; BChE = 7.00 μM).
  • Molecular formula C18H11ClO3; molecular weight 310.73 g/mol.
  • High purity (99.05% by supplier analysis).
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for enzymatic assays, inhibitor screening, and cell-based studies.
  • Research use only; not for human therapeutic use.
Catalog No. 50-002-63156 Supplier Medchemexpress LLC Supplier No. HY1329075MG
MAO-B-IN-2 is a small-molecule, selective and competitive inhibitor of monoamine oxidase B (MAO-B) and butyrylcholinesterase (BChE). It is provided for research use and is available as a solid and as a 10 mM solution in DMSO for biochemical and cell-based studies.
  • Selective MAO-B and BChE inhibition (IC50 MAO-B = 0.51 μM; BChE = 7.00 μM).
  • Molecular formula C18H11ClO3; molecular weight 310.73 g/mol.
  • High purity (99.05% by supplier analysis).
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for enzymatic assays, inhibitor screening, and cell-based studies.
  • Research use only; not for human therapeutic use.