Learn More
Medchemexpress LLC (E)-3-(3-(2-chlorophenyl)acryloyl)-2H-chromen-2-one | 1253978-24-3 | 99.1% | C18H11ClO3 | 10MG
SDP
Catalog No. 5000259368
Encompass_Preferred
MAO-B-IN-2 is a selective, competitive inhibitor of monoamine oxidase B (MAO-B) and butyrylcholinesterase (BChE) prepared for research use. It shows low-micromolar potency against MAO-B and moderate activity against BChE, is supplied as a characterized solid with high purity, and includes recommended storage guidelines for stability.
Catalog No. 50-002-59368 Supplier Medchemexpress LLC Supplier No. HY13290710MG
MAO-B-IN-2 is a selective, competitive inhibitor of monoamine oxidase B (MAO-B) and butyrylcholinesterase (BChE) prepared for research use. It shows low-micromolar potency against MAO-B and moderate activity against BChE, is supplied as a characterized solid with high purity, and includes recommended storage guidelines for stability.