Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3-(3-phenyl-2,1-benzoxazol-5-yl)prop-2-enoic acid | 2967648-66-2 | 99.7% | 5 MG
SDP

Catalog No. 5000230210
Encompass_Preferred

IP6K2-IN-2 is a small-molecule inhibitor of inositol hexakisphosphate kinase 2 (IP6K2) with submicromolar activity. It inhibits IP6K2 (IC50 0.58 μM), IP6K1 (IC50 0.86 μM), and IP6K3 (IC50 3.08 μM). The compound is supplied with high purity and is used as a research tool in studies of metabolic regulation and obesity.

  • Selective inhibitor of IP6K2 with submicromolar potency.
  • Reported activity against related isoforms IP6K1 and IP6K3.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO (approx. 25 mg/mL) for solution-based assays.
  • Available as solid and as prepared DMSO solution for assay flexibility.
  • Storage recommendations: powder at -20°C; solutions at -80°C for long-term stability.
  • Intended for research use in metabolic and obesity-related studies.

Catalog No. 50-002-30210 Supplier Medchemexpress LLC Supplier No. HY122706A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IP6K2-IN-2 is a small-molecule inhibitor of inositol hexakisphosphate kinase 2 (IP6K2) with submicromolar activity. It inhibits IP6K2 (IC50 0.58 μM), IP6K1 (IC50 0.86 μM), and IP6K3 (IC50 3.08 μM). The compound is supplied with high purity and is used as a research tool in studies of metabolic regulation and obesity.

  • Selective inhibitor of IP6K2 with submicromolar potency.
  • Reported activity against related isoforms IP6K1 and IP6K3.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO (approx. 25 mg/mL) for solution-based assays.
  • Available as solid and as prepared DMSO solution for assay flexibility.
  • Storage recommendations: powder at -20°C; solutions at -80°C for long-term stability.
  • Intended for research use in metabolic and obesity-related studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.