Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3-[4,5-bis(1,3-benzodioxol-5-yl)furan-2-yl]prop-2-enoic acid | 2502294-55-3 | 99.7% | 378.33 | 50 MG
SDP

Catalog No. 5000362267
Encompass_Preferred

OSM-SMI-10B is a derivative of OSM-SMI-10 that significantly reduces Oncostatin M (OSM)-induced STAT3 phosphorylation in cancer cells upon co-incubation with OSM.

  • Targets p-STAT3
  • Molecular weight: 378.33
  • Formula: C21H14O7
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in vitro: DMSO: 250 mg/mL (660.80 mM)
  • Solubility in vivo: ≥ 6.25 mg/mL (16.52 mM)

Catalog No. 50-003-62267 Supplier Medchemexpress LLC Supplier No. HY14869250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OSM-SMI-10B is a derivative of OSM-SMI-10 that significantly reduces Oncostatin M (OSM)-induced STAT3 phosphorylation in cancer cells upon co-incubation with OSM.

  • Targets p-STAT3
  • Molecular weight: 378.33
  • Formula: C21H14O7
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Solubility in vitro: DMSO: 250 mg/mL (660.80 mM)
  • Solubility in vivo: ≥ 6.25 mg/mL (16.52 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.