Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3,4,5-trimethoxycinnamic acid | 20329-98-0 | 99.95% | 238.24 g/mol | C12H14O5 | 100 MG
SDP

Catalog No. 5000209765
Encompass_Preferred

(E)-3,4,5-Trimethoxycinnamic acid is a cinnamic acid derivative used as a research reagent in pharmacology and biochemical studies, including investigations of GABAA/BZ receptor modulation and anticonvulsant or sedative effects. It is supplied as a solid powder for in vitro and in vivo research and should be handled according to institutional safety procedures.

  • High reported purity suitable for analytical and biological studies.
  • Molecular weight 238.24 g/mol and formula C12H14O5.
  • Available in multiple pack sizes, including 100 MG for small-scale experiments.
  • Storage guidance provided for powder and solutions to preserve stability.
  • Certificate of analysis and product data sheet available.

Catalog No. 50-002-09765 Supplier Medchemexpress LLC Supplier No. HYW050162100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3,4,5-Trimethoxycinnamic acid is a cinnamic acid derivative used as a research reagent in pharmacology and biochemical studies, including investigations of GABAA/BZ receptor modulation and anticonvulsant or sedative effects. It is supplied as a solid powder for in vitro and in vivo research and should be handled according to institutional safety procedures.

  • High reported purity suitable for analytical and biological studies.
  • Molecular weight 238.24 g/mol and formula C12H14O5.
  • Available in multiple pack sizes, including 100 MG for small-scale experiments.
  • Storage guidance provided for powder and solutions to preserve stability.
  • Certificate of analysis and product data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.