Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3,4,5-trimethoxycinnamic acid | 20329-98-0 | MFCD00004388 | 100.0% | 238.24 g/mol | C12H14O5 | 500 MG
SDP

Supplier:  Medchemexpress LLC HYW050162500MG

Encompass_Preferred

(E)-3,4,5-trimethoxycinnamic acid is a purified small-molecule research reagent used in biochemical and pharmacological studies, including investigation of GABAA/BZ receptor activity. It is supplied as a solid with a reported molecular weight of 238.24 g/mol and high purity for in vitro applications.

  • Identifies as (E)-3,4,5-trimethoxycinnamic acid (CAS 20329-98-0).
  • Molecular formula: C12H14O5.
  • Molecular weight: 238.24 g/mol.
  • High reported purity suitable for analytical and biological assays.
  • Available in multiple pack sizes for flexible experimental use.

Catalog No. 50-002-10295


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.