Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(E)-3,4,5-Trimethoxycinnamic Acid 98.0+%, TCI America™
SDP

Catalog No. T110425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T110425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T110425G Supplier TCI America Supplier No. T110425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20329-98-0
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.24
MDL Number MFCD00004388
InChI Key YTFVRYKNXDADBI-UHFFFAOYSA-N
Synonym (E)-3-(3,4,5-Trimethoxyphenyl)acrylic Acid
PubChem CID 735755
IUPAC Name 3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(C=CC(O)=O)=CC(OC)=C1OC

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 25 g
Formula Weight 238.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (E)-3,4,5-Trimethoxycinnamic Acid
EINECSNumber (3)-4314
RTECSNumber GE0722000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.