Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-(3-(4-bromo-2-hydroxyphenyl)-2-((5-methyl-1H-benzo[d]imidazol-2-yl)thio)acryloyl)hydrazine | 330676-02-3 | MFCD01230899 | 99.0% | 419.30 | C17H15BrN4O2S | 100 MG
SDP

Catalog No. 5000246670
Encompass_Preferred

KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor and cAMP pathway inhibitor intended for research use. It inhibits recombinant human sACt and heterologously expressed sACt in cellular assays (IC50 ~3-10 μM), and is used in biochemical and cell-based studies of cAMP signaling.

  • High purity (99.0%) suitable for biochemical assays.
  • Molecular weight 419.30; formula C17H15BrN4O2S.
  • CAS number 330676-02-3 for unambiguous identification.
  • Available as a solid 100 mg package and as a 10 mM, 1 mL solution in DMSO.
  • For research use only; not for clinical or human applications.

Catalog No. 50-002-46670 Supplier Medchemexpress LLC Supplier No. HY103194100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor and cAMP pathway inhibitor intended for research use. It inhibits recombinant human sACt and heterologously expressed sACt in cellular assays (IC50 ~3-10 μM), and is used in biochemical and cell-based studies of cAMP signaling.

  • High purity (99.0%) suitable for biochemical assays.
  • Molecular weight 419.30; formula C17H15BrN4O2S.
  • CAS number 330676-02-3 for unambiguous identification.
  • Available as a solid 100 mg package and as a 10 mM, 1 mL solution in DMSO.
  • For research use only; not for clinical or human applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.