Learn More
Medchemexpress LLC (E)-3-(4-chlorobenzyl)-2-(2-(pyridin-3-yl)vinyl)quinazolin-4(3H)-one | 2101739-18-6 | 99.9% | 10 MG
SDP
Catalog No. 5000231465
Encompass_Preferred
RAD51-IN-1 is a small-molecule inhibitor of the RAD51 recombinase, described as a derivative of B02 and used in preclinical cancer research to study homologous recombination and DNA repair pathways.
  • Potent RAD51 inhibitor for studying DNA repair
  • Molecular formula C22H16ClN3O and molecular weight 373.83
  • High purity suitable for biochemical assays (listed as 99.89%)
  • Available in multiple package sizes, including 10 MG
  • Yellow to orange solid, commonly supplied as a research reagent
Catalog No. 50-002-31465 Supplier Medchemexpress LLC Supplier No. HY12270510MG
RAD51-IN-1 is a small-molecule inhibitor of the RAD51 recombinase, described as a derivative of B02 and used in preclinical cancer research to study homologous recombination and DNA repair pathways.
  • Potent RAD51 inhibitor for studying DNA repair
  • Molecular formula C22H16ClN3O and molecular weight 373.83
  • High purity suitable for biochemical assays (listed as 99.89%)
  • Available in multiple package sizes, including 10 MG
  • Yellow to orange solid, commonly supplied as a research reagent