Learn More
Medchemexpress LLC (E)-3,4-Dimethoxycinnamic acid | 14737-89-4 | 208.21 | 50 G
SDP
Catalog No. 5000415192
Encompass_Preferred
(E)-3,4-Dimethoxycinnamic acid is the less active isomer of 3,4-Dimethoxycinnamic acid. This compound exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway and is intended for research use only.
  • Attenuates ethanol-induced apoptosis of ethanol-exposed L-02 cells.
  • Inhibits the expression levels of Nox4 and p22phox in L-02 cells.
  • Attenuates ethanol-induced MAPK phosphorylation in L-02 cells.
  • Decreases the expression levels of superoxide dismutase, catalase, and phospholipid hydroperoxide gluthione peroxidase.
  • Downregulates the levels of Bax/Bcl-2 and the cleaved forms of caspase-3.
Catalog No. 50-004-15192 Supplier Medchemexpress LLC Supplier No. HYN1778A50G
Add to Cart
Add to Cart
(E)-3,4-Dimethoxycinnamic acid is the less active isomer of 3,4-Dimethoxycinnamic acid. This compound exerts anti-apoptotic effects on L-02 cells via the ROS-mediated signaling pathway and is intended for research use only.
  • Attenuates ethanol-induced apoptosis of ethanol-exposed L-02 cells.
  • Inhibits the expression levels of Nox4 and p22phox in L-02 cells.
  • Attenuates ethanol-induced MAPK phosphorylation in L-02 cells.
  • Decreases the expression levels of superoxide dismutase, catalase, and phospholipid hydroperoxide gluthione peroxidase.
  • Downregulates the levels of Bax/Bcl-2 and the cleaved forms of caspase-3.