Learn More
Medchemexpress LLC HY-W068771 500mg Medchemexpress, (E)-3-(4-Methoxyphenyl)acrylic acid CAS:943-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW068771500MG
Encompass_Preferred
Medchemexpress, HY-W068771 500mg (E)-3-(4-Methoxyphenyl)acrylic acid CAS:943-89-5 (E)-3-(4-Methoxyphenyl)acrylic acid (compound 3) is isolated from Arachis hypogaea, Scrophularia buergeriana Miquel, Aquilegia vulgaris, Anigozanthos preissii and so on. (E)-3-(4-Methoxyphenyl)acrylic acid shows significant hepatoprotective activity, anti-amnesic,, cognition-enhancing activity, antihyperglycemic, and neuroprotective activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3502