Learn More
Medchemexpress LLC (E)-3-(4-methoxyphenyl)acrylic acid | 943-89-5 | MFCD00004398 | 178.18 g/mol | C10H10O3 | 1 ML
SDP
Catalog No. 5000210817
Encompass_Preferred
(E)-3-(4-methoxyphenyl)acrylic acid (CAS 943-89-5) is provided as a ready-to-use 10 mM solution in DMSO for research use. The compound is commonly used as a reference standard in metabolism-related studies and should be stored according to SDS recommendations to preserve stability.
  • Ready-to-use 10 mM solution in DMSO.
  • Suitable as a reference compound for metabolism research.
  • Molecular formula C10H10O3, molecular weight 178.18 g/mol.
  • Store according to SDS: powdered and in-solvent temperature recommendations to maintain stability.
Catalog No. 50-002-10817 Supplier Medchemexpress LLC Supplier No. HYW06877110MM/1ML
(E)-3-(4-methoxyphenyl)acrylic acid (CAS 943-89-5) is provided as a ready-to-use 10 mM solution in DMSO for research use. The compound is commonly used as a reference standard in metabolism-related studies and should be stored according to SDS recommendations to preserve stability.
  • Ready-to-use 10 mM solution in DMSO.
  • Suitable as a reference compound for metabolism research.
  • Molecular formula C10H10O3, molecular weight 178.18 g/mol.
  • Store according to SDS: powdered and in-solvent temperature recommendations to maintain stability.