Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3-(4-Methoxyphenyl)acrylic acid | 943-89-5 | MFCD00004398 | 99.7% | 178.18 g/mol | C10H10O3 | 25 G
SDP

Catalog No. 5000209537
Encompass_Preferred

(E)-3-(4-Methoxyphenyl)acrylic acid is the E-isomer of 3-(4-methoxyphenyl)acrylic acid supplied as a solid reference compound for research and analytical use. It is reported with high purity and has supporting documentation for safety and quality verification.

  • Commercially available as a solid in multiple pack sizes.
  • Reported purity of approximately 99.7%.
  • Molecular formula C10H10O3.
  • Molecular weight 178.18 g/mol.
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-09537 Supplier Medchemexpress LLC Supplier No. 25GHYW068771
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3-(4-Methoxyphenyl)acrylic acid is the E-isomer of 3-(4-methoxyphenyl)acrylic acid supplied as a solid reference compound for research and analytical use. It is reported with high purity and has supporting documentation for safety and quality verification.

  • Commercially available as a solid in multiple pack sizes.
  • Reported purity of approximately 99.7%.
  • Molecular formula C10H10O3.
  • Molecular weight 178.18 g/mol.
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.