Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3,4-(methylenedioxy)cinnamic acid | 38489-76-8 | MFCD00005837 | 99.9% | 192.17 g/mol | C10H8O4 | 10 MG
SDP

Catalog No. 5000210825
Encompass_Preferred

(E)-3,4-(Methylenedioxy)cinnamic acid is a cinnamic acid derivative obtained from the stem bark of Brombya platynema. It is supplied for research use only and is characterized by defined physicochemical properties.

  • Molecular formula: C10H8O4.
  • Molecular weight: 192.17 g/mol.
  • CAS number: 38489-76-8.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Storage: store at -20°C and protect from light; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Synonyms: (E)-piperonylprop-2-enoic acid.

Catalog No. 50-002-10825 Supplier Medchemexpress LLC Supplier No. HYW06878310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3,4-(Methylenedioxy)cinnamic acid is a cinnamic acid derivative obtained from the stem bark of Brombya platynema. It is supplied for research use only and is characterized by defined physicochemical properties.

  • Molecular formula: C10H8O4.
  • Molecular weight: 192.17 g/mol.
  • CAS number: 38489-76-8.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Storage: store at -20°C and protect from light; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Synonyms: (E)-piperonylprop-2-enoic acid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.