Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3',6-Disinapoylsucrose | 139891-98-8 | 754.69 | 5 MG
SDP

Catalog No. 5000414667
Encompass_Preferred

(E)-3',6-Disinapoylsucrose is the index component of Yuanzhi (Polygala tenuifolia Willd), exhibiting potent antioxidant activity and antidepressant effects. This solid compound is derived from plants and is white to yellow in appearance.

  • Purity: 99.59%
  • Appearance: Solid
  • Color: White to yellow
  • Initial source: Plants, Polygalaceae, Polygala tenuifolia Willd.

Catalog No. 50-004-14667 Supplier Medchemexpress LLC Supplier No. HYN14145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3',6-Disinapoylsucrose is the index component of Yuanzhi (Polygala tenuifolia Willd), exhibiting potent antioxidant activity and antidepressant effects. This solid compound is derived from plants and is white to yellow in appearance.

  • Purity: 99.59%
  • Appearance: Solid
  • Color: White to yellow
  • Initial source: Plants, Polygalaceae, Polygala tenuifolia Willd.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.