Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3',6-disinapoylsucrose | 139891-98-8 | MFCD23379921 | 99.6% | 754.69 g/mol | C34H42O19 | 10 MG
SDP

Catalog No. 5000295742
Encompass_Preferred

(E)-3',6-disinapoylsucrose is a natural oligosaccharide ester isolated from Polygala tenuifolia and supplied as a high-purity analytical research standard. It exhibits antioxidant and antidepressant activities and is intended for laboratory research use only.

  • High purity, 99.6%.
  • Molecular weight 754.69 g/mol.
  • CAS number 139891-98-8.
  • Suitable for analytical, pharmacological, and biochemical research.
  • Provided in small, accurately weighed pack sizes for analytical workflows.
  • Intended for research use only; not for human consumption.

Catalog No. 50-002-95742 Supplier Medchemexpress LLC Supplier No. HYN141410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3',6-disinapoylsucrose is a natural oligosaccharide ester isolated from Polygala tenuifolia and supplied as a high-purity analytical research standard. It exhibits antioxidant and antidepressant activities and is intended for laboratory research use only.

  • High purity, 99.6%.
  • Molecular weight 754.69 g/mol.
  • CAS number 139891-98-8.
  • Suitable for analytical, pharmacological, and biochemical research.
  • Provided in small, accurately weighed pack sizes for analytical workflows.
  • Intended for research use only; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.