Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [(E)-[(3-bromophenyl)-(3-hydroxyphenyl)methylidene]amino]thiourea | 1131456-28-4 | 99.8% | 350.23 g/mol | C14H12BrN3OS | 10 MG
SDP

Catalog No. 5000228066
Encompass_Preferred

KGP94 is a selective cathepsin L inhibitor used in research to inhibit migration and invasion of metastatic carcinoma cells. It shows potent biochemical activity (IC50 ≈ 189 nM) and low cytotoxicity (GI50 ≈ 26.9 μM). Supplied as a high-purity solid for biochemical and cellular studies.

  • Molecular formula: C14H12BrN3OS.
  • Molecular weight: 350.23 g/mol.
  • Cas number: 1131456-28-4.
  • Purity: 99.8% (reported).
  • Solubility: highly soluble in DMSO; in vivo formulations reported.
  • Storage: powder -20°C for long term; solutions stored at -80°C.

Catalog No. 50-002-28066 Supplier Medchemexpress LLC Supplier No. HY11876210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KGP94 is a selective cathepsin L inhibitor used in research to inhibit migration and invasion of metastatic carcinoma cells. It shows potent biochemical activity (IC50 ≈ 189 nM) and low cytotoxicity (GI50 ≈ 26.9 μM). Supplied as a high-purity solid for biochemical and cellular studies.

  • Molecular formula: C14H12BrN3OS.
  • Molecular weight: 350.23 g/mol.
  • Cas number: 1131456-28-4.
  • Purity: 99.8% (reported).
  • Solubility: highly soluble in DMSO; in vivo formulations reported.
  • Storage: powder -20°C for long term; solutions stored at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.