Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (E)-3-(Diethylamino)-1-(4-fluoro-2-hydroxyphenyl)prop-2-en-1-one

Catalog No. JRD08481G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD08481G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD08481G Supplier Sigma Aldrich Supplier No. JRD08481G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pentafluorophenyl isocyanate (pfpi), an aryl isocyanate, is an n-protecting reagent. the photoinduced intermolecular carbamoylation of tertiary ethereal c-h bond of fused bicyclic system using pfpi/benzophenone has been reported. reacting wood polymers with pfpi can make them resistant to fungal attack by forming a stable carbamate bond.pentafluorophenyl isocyanate may be used in the preparation of:
  • Pentafluorophenyl end-capped poly(ethylene glycol) (pf-peg-pf)
  • 9-o-(pentafluorophenylcarbamoyl)quinine
  • 9-o-(tert-butyldimethylsilyl)-10-(pentafluorophenylcarbamoyloxy)-6α-methoxy-11-norcinchonan-9-ol
  • Substituted 2,6-ethynylpyridine bisphenylurea scaffolds
  • Novel o-xylyl bridged hexaurea receptor for atmospheric co2 uptake

Specifications

Molecular Formula C13H16FNO2
Linear Formula C13H16FNO2
MDL Number MFCD12196361
Quantity 5 mL
Molecular Weight (g/mol) 237.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.