Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3-pentenoic acid | 1617-32-9 | MFCD00063291 | ≥95.0% | 100.12 g·mol-1 | C5H8O2 | 1 G
SDP

Catalog No. 5000208362
Encompass_Preferred

(E)-3-pentenoic acid is an unsaturated short-chain carboxylic acid supplied as a research reagent and synthetic intermediate for organic chemistry and biochemical studies. It is provided as a high-purity material intended for laboratory research and not for human use.

  • Unsaturated carboxylic acid with formula C5H8O2.
  • Molecular weight 100.12 g·mol-1.
  • Purity ≥95.0%.
  • Available in small pack sizes suitable for research, for example 1 g.
  • Commonly used as a building block and intermediate in organic synthesis.
  • Intended for research use only; not for human or therapeutic applications.

Catalog No. 50-002-08362 Supplier Medchemexpress LLC Supplier No. HYW0885011G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3-pentenoic acid is an unsaturated short-chain carboxylic acid supplied as a research reagent and synthetic intermediate for organic chemistry and biochemical studies. It is provided as a high-purity material intended for laboratory research and not for human use.

  • Unsaturated carboxylic acid with formula C5H8O2.
  • Molecular weight 100.12 g·mol-1.
  • Purity ≥95.0%.
  • Available in small pack sizes suitable for research, for example 1 g.
  • Commonly used as a building block and intermediate in organic synthesis.
  • Intended for research use only; not for human or therapeutic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.