Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-3-pentenoic acid | 1617-32-9 | MFCD00063291 | ≥95.0% | 100.12 g·mol⁻¹ | C5H8O2 | 250 MG
SDP

Catalog No. 5000208480
Encompass_Preferred

(E)-3-pentenoic acid is an unsaturated carboxylic acid supplied for laboratory research and analytical use. The material is a liquid, colorless to light yellow, and is provided with supporting documentation for safe handling and quality verification.

  • High purity (≥95.0%).
  • Molecular weight 100.12 g·mol⁻¹.
  • Molecular formula C5H8O2.
  • Available in small laboratory quantities including 250 MG.
  • Provided with SDS and certificate of analysis for traceability.

Catalog No. 50-002-08480 Supplier Medchemexpress LLC Supplier No. 250MGHYW088501
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-3-pentenoic acid is an unsaturated carboxylic acid supplied for laboratory research and analytical use. The material is a liquid, colorless to light yellow, and is provided with supporting documentation for safe handling and quality verification.

  • High purity (≥95.0%).
  • Molecular weight 100.12 g·mol⁻¹.
  • Molecular formula C5H8O2.
  • Available in small laboratory quantities including 250 MG.
  • Provided with SDS and certificate of analysis for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.