Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (E)-3-(Tert-Butyldimethylsilyloxy)Propene-1-Yl-Boronic Acid Pinacol Ester

Catalog No. 7102105G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-021-05G 5 g
71-021-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-021-05G Supplier Sigma Aldrich Supplier No. 7102105G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(E)-3-(tert-Butyldimethylsilyloxy)propene-1-yl-boronic acid pinacol ester can be used as a reactant in: Palladium-catalyzed Suzuki-Miyaura coupling reaction. The synthesis of anti-homoallylic alcohols by allylation reaction with aldehydes in the presence of iridium complex-catalyst. The preparation of +a-imino aldehydes.

Specifications

CAS 114653-19-9
Density 0.910 g/mL at 25°C
Molecular Formula C15H31BO3Si
Linear Formula C15H31BO3Si
MDL Number MFCD15144803
Quantity 5 g
Synonym (E)-2-[3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1-propenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 2-[(1E)-3-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1-propenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; tert-Butyldimethyl[[(2E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-en-1-yl]oxy]silane
Molecular Weight (g/mol) 298.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.