Learn More
Miele, Inc. E 353 Mielava Clip for use with E 351 Injector
Catalog No. 501713009
Encompass_Preferred
E 353 Clamps
Height-adjustable for optimum positioning of laboratory glassware on the nozzle.
Suitable for injector nozzle E 351
Height adjustable locking
Suitable for bottles, Erlenmeyer flasks and round bottomed flasks for example
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3009 Supplier Miele, Inc. Supplier No. 03809530
E 353 Clamps
Height-adjustable for optimum positioning of laboratory glassware on the nozzle.
Suitable for injector nozzle E 351
Height adjustable locking
Suitable for bottles, Erlenmeyer flasks and round bottomed flasks for example
Visit www.mieleusa.com/professional for technical information.