Learn More
Miele, Inc. E 354 Mielava clip for use with E 352 injector
Catalog No. 501713011
Encompass_Preferred
E 354 Clamps
Height-adjustable for optimum positioning of laboratory glassware on the nozzle.
Suitable for injector nozzle E 352
Height adjustable locking
Suitable for bottles, Erlenmeyer flasks and round bottomed flasks for example
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3011 Supplier Miele, Inc. Supplier No. 03809540
Add to Cart
Add to Cart
E 354 Clamps
Height-adjustable for optimum positioning of laboratory glassware on the nozzle.
Suitable for injector nozzle E 352
Height adjustable locking
Suitable for bottles, Erlenmeyer flasks and round bottomed flasks for example
Visit www.mieleusa.com/professional for technical information.