Learn More
Medchemexpress LLC E-3620 | 151213-86-4 | 98.88% | 412.35 | 250 MG
SDP
Catalog No. 5000385315
Encompass_Preferred
E-3620 is a potent 5-HT3 receptor antagonist. It can be utilized for research related to dyskinesia and gastrointestinal motility.
  • Potent 5-HT3 receptor antagonist.
  • Used in research for dyskinesia.
  • Used in research for gastrointestinal motility.
Catalog No. 50-003-85315 Supplier Medchemexpress LLC Supplier No. 250MGHY19199
E-3620 is a potent 5-HT3 receptor antagonist. It can be utilized for research related to dyskinesia and gastrointestinal motility.
  • Potent 5-HT3 receptor antagonist.
  • Used in research for dyskinesia.
  • Used in research for gastrointestinal motility.