Learn More
Medchemexpress LLC (E)-(4-(2-(1H-indazol-3-yl)vinyl)phenyl)(piperazin-1-yl)methanone | 1000669-72-6 | MFCD18385006 | 99.9% | 332.40 g/mol | C20H20N4O | 50 MG
SDP
Catalog No. 5000244721
Encompass_Preferred
KW-2449 is a small-molecule, multi-targeted kinase inhibitor used in preclinical oncology research to study FLT3- and BCR-ABL-driven leukemia models. It inhibits FLT3, ABL (including the T315I mutant), and Aurora kinases, and is characterized by a molecular formula of C20H20N4O and high chemical purity.
Catalog No. 50-002-44721 Supplier Medchemexpress LLC Supplier No. HY1033950MG
KW-2449 is a small-molecule, multi-targeted kinase inhibitor used in preclinical oncology research to study FLT3- and BCR-ABL-driven leukemia models. It inhibits FLT3, ABL (including the T315I mutant), and Aurora kinases, and is characterized by a molecular formula of C20H20N4O and high chemical purity.