Learn More
Medchemexpress LLC (E)-4-(2-chloro-6-fluorostyryl)-N-methylaniline | 1391934-98-7 | 98.5% | C15H13ClFN | 10MG
SDP
Catalog No. 5000261210
Encompass_Preferred
FIDAS-5 is a small-molecule research compound that inhibits methionine S-adenosyltransferase 2A (MAT2A). It competitively displaces S-adenosylmethionine with a reported IC50 of 2.1 μM and has demonstrated anticancer activity in preclinical studies. The material is supplied as a high-purity solid powder for laboratory research and is unstable in solution, so fresh preparations are recommended.
  • Potent MAT2A inhibitor with IC50 = 2.1 μM.
  • High purity (≈98.5%).
  • Molecular formula C15H13ClFN; molecular weight 261.72.
  • Supplied as a solid; available in small pack sizes including 10 mg.
  • Powder stable at -20°C; unstable in solution, prepare fresh solutions.
  • Used in preclinical and in vitro anticancer research.
Catalog No. 50-002-61210 Supplier Medchemexpress LLC Supplier No. HY13614410MG
FIDAS-5 is a small-molecule research compound that inhibits methionine S-adenosyltransferase 2A (MAT2A). It competitively displaces S-adenosylmethionine with a reported IC50 of 2.1 μM and has demonstrated anticancer activity in preclinical studies. The material is supplied as a high-purity solid powder for laboratory research and is unstable in solution, so fresh preparations are recommended.
  • Potent MAT2A inhibitor with IC50 = 2.1 μM.
  • High purity (≈98.5%).
  • Molecular formula C15H13ClFN; molecular weight 261.72.
  • Supplied as a solid; available in small pack sizes including 10 mg.
  • Powder stable at -20°C; unstable in solution, prepare fresh solutions.
  • Used in preclinical and in vitro anticancer research.