Learn More
Medchemexpress LLC (E)-4-(2-Nitrophenyl)-4-oxobut-2-enoic acid | 80937-24-2 | 95.0% | 221.17 | 100 MG
SDP
Catalog No. 5000403733
Encompass_Preferred
(E)-4-(2-Nitrophenyl)-4-oxobut-2-enoic acid is an intermediate chemical compound. It is used to synthesize Phenyl-4-oxo-2-butenoic acid, which exhibits high antiulcer and antisecretory activities.
  • Can be used to synthesize Phenyl-4-oxo-2-butenoic acid
  • Phenyl-4-oxo-2-butenoic acid has high antiulcer activity
  • Phenyl-4-oxo-2-butenoic acid has high antisecretory activity
Catalog No. 50-004-03733 Supplier Medchemexpress LLC Supplier No. HY400142100MG
(E)-4-(2-Nitrophenyl)-4-oxobut-2-enoic acid is an intermediate chemical compound. It is used to synthesize Phenyl-4-oxo-2-butenoic acid, which exhibits high antiulcer and antisecretory activities.
  • Can be used to synthesize Phenyl-4-oxo-2-butenoic acid
  • Phenyl-4-oxo-2-butenoic acid has high antiulcer activity
  • Phenyl-4-oxo-2-butenoic acid has high antisecretory activity