Learn More
Medchemexpress LLC (E)-4-carboxystyryl-4-chlorobenzylsulfone | 334969-03-8 | MFCD07366276 | 99.9% | 336.8 g/mol | C16H13ClO4S | 100 MG
SDP
Catalog No. 5000248983
Encompass_Preferred
Recilisib is a small-molecule radioprotectant that mitigates radiation-induced damage by up-regulating the PI3K/AKT cell-survival signaling cascade. Supplied as a white to off-white solid for laboratory research, the compound has been characterized in preclinical studies and is distributed for investigational use with high reported purity.
  • Radioprotective activity through PI3K/AKT pathway activation.
  • High purity suitable for research assays (≈99.9%).
  • Solid form, soluble in dimethyl sulfoxide for experimental use.
  • Available in laboratory quantities for preclinical studies (e.g., 100 mg).
  • Intended for research use only; not for human use.
Catalog No. 50-002-48983 Supplier Medchemexpress LLC Supplier No. HY101625100MG
Recilisib is a small-molecule radioprotectant that mitigates radiation-induced damage by up-regulating the PI3K/AKT cell-survival signaling cascade. Supplied as a white to off-white solid for laboratory research, the compound has been characterized in preclinical studies and is distributed for investigational use with high reported purity.
  • Radioprotective activity through PI3K/AKT pathway activation.
  • High purity suitable for research assays (≈99.9%).
  • Solid form, soluble in dimethyl sulfoxide for experimental use.
  • Available in laboratory quantities for preclinical studies (e.g., 100 mg).
  • Intended for research use only; not for human use.