Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (E)-4-Chloronicotinaldehyde oxime

Catalog No. ADE0006611G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0006611G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0006611G Supplier Sigma Aldrich Supplier No. ADE0006611G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Precursor of the unstable methyl 3-oxo-4-pentenoate obtained by acid catalysis (e.g
  • Tosoh) used for annulation reactions
  • It can be prepared in-situ, review

Specifications

CAS 471909-43-0
Molecular Formula C6H5ClN2O
Linear Formula C6H5ClN2O
MDL Number MFCD12922730
Quantity 5 mL
Molecular Weight (g/mol) 156.57
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.