Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-4-Hydroxytamoxifen | 174592-47-3 | 98.3% | 387.51 | 1 ML
SDP

Catalog No. 5000390537
Encompass_Preferred

(E)-4-Hydroxytamoxifen ((E)-Afimoxifene), the less active isomer of (Z)-4-hydroxytamoxifen, is an estrogen receptor modulator.

  • Purity of 98.3%
  • Specific CAS number: 174592-47-3
  • Appears as a white to off-white solid
  • Functions as an estrogen receptor modulator
  • Features specific storage conditions for both powder and solution forms
  • Soluble in DMSO (12.5 mg/mL)

Catalog No. 50-003-90537 Supplier Medchemexpress LLC Supplier No. HY16950B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(E)-4-Hydroxytamoxifen ((E)-Afimoxifene), the less active isomer of (Z)-4-hydroxytamoxifen, is an estrogen receptor modulator.

  • Purity of 98.3%
  • Specific CAS number: 174592-47-3
  • Appears as a white to off-white solid
  • Functions as an estrogen receptor modulator
  • Features specific storage conditions for both powder and solution forms
  • Soluble in DMSO (12.5 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.