Learn More
Medchemexpress LLC (E)-4-(Naphthalen-2-yl)but-3-en-2-one | 51557-10-9 | 99.9% | 196.24 | 250 MG
SDP
Catalog No. 5000402694
Encompass_Preferred
(E)-4-(Naphthalen-2-yl)but-3-en-2-one is a small active molecule that can be used as a building block. It exhibits anti-leukemic activity, inhibiting the proliferation of human chronic myeloid leukemia cell K562 with an IC50 of 7.6 μM.
  • Can be used as a building block.
  • Exhibits anti-leukemic activity.
  • Inhibits the proliferation of human chronic myeloid leukemia cell K562.
Catalog No. 50-004-02694 Supplier Medchemexpress LLC Supplier No. 250MGHY402815
(E)-4-(Naphthalen-2-yl)but-3-en-2-one is a small active molecule that can be used as a building block. It exhibits anti-leukemic activity, inhibiting the proliferation of human chronic myeloid leukemia cell K562 with an IC50 of 7.6 μM.
  • Can be used as a building block.
  • Exhibits anti-leukemic activity.
  • Inhibits the proliferation of human chronic myeloid leukemia cell K562.