Learn More
Miele, Inc. E 402 Half Insert for 44 watch-glasses (Ø 80-125 mm)
Catalog No. 501713022
Encompass_Preferred
E 402 Mesh Basket
For optimum loading of up to 44 watch glasses.
Suitable for 44 watch glasses Ø 80 - 125 mm
Equipped with 23 bars at a distance of 0 5/8 (15) mm
Visit www.mieleusa.com/professional for technical information.
Catalog No. 50-171-3022 Supplier Miele, Inc. Supplier No. 03830420
E 402 Mesh Basket
For optimum loading of up to 44 watch glasses.
Suitable for 44 watch glasses Ø 80 - 125 mm
Equipped with 23 bars at a distance of 0 5/8 (15) mm
Visit www.mieleusa.com/professional for technical information.